Documenti fondamentali
WHA10370381
Whatman® glass microfiber filters with organic binder, Grade GF 10
roll, W × L 30 mm × 20 m, pack of 1
Sinonimo/i:
fiber filters, glass filter, whatman filter
About This Item
Codice UNSPSC:
40161507
NACRES:
NB.24
Materiali
glass membrane (with organic binder)
roll
Sterilità
non-sterile
Confezionamento
pack of 1
Produttore/marchio commerciale
Whatman 10370381
Whatman Article No. 28414797 (US reference)
Largh. × lungh.
30 mm × 20 m
Descrizione generale
This filter with extreme mechanical stability and temperature resistant up to 180°C is used as a weighing aid for infrared weighing and as a roll filter in automatic air filtration units.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)