Documenti fondamentali
T308579
7,8-DIETHYL-10-(1'-D-RIBITYL)ISOALLOXAZINE
AldrichCPR
Scegli un formato
T308579-100MG
158,00 €
158,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
T308579-100MG
158,00 €
About This Item
Formula condensata:
C19H24N4O6
Numero CAS:
Peso molecolare:
404.43
Numero MDL:
158,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Altre note
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)