Documenti fondamentali
S434795
3-AMINO-2,5-DICHLOROBENZOIC ACID, SODIUM SALT, TECH.
AldrichCPR
Scegli un formato
S434795-1G
29,00 €
29,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
S434795-1G
29,00 €
About This Item
Formula condensata:
C7H4Cl2NNaO2
Numero CAS:
Peso molecolare:
228.011
Numero MDL:
29,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Altre note
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)