Documenti
GF36594838
Alumina
foam, thickness 12.7 mm, size 100 × 100 mm, porosity 0.84
Sinonimo/i:
AL603825
About This Item
Codice UNSPSC:
11181511
NACRES:
NA.23
Produttore/marchio commerciale
Goodfellow 365-948-38
Misura
100 × 100 mm
Spessore
12.7 mm