CLS3606060F
Pyrex® Buchner funnel with fritted disc
capacity 60 mL, porosity 4-5.5 μm
Sinonimo/i:
Corning® 36060
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Prodotti consigliati
Produttore/marchio commerciale
Corning 36060-60F
Capacità
60 mL
Diam. disco
40 mm
Dimensione pori
4-5.5 μm porosity
Descrizione generale
Especially useful where filter paper would be attacked by filtered material. Disc should not be subjected to pressure differential in excess of 1 atm. Top edge is beaded. Granular frit.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)