Documenti fondamentali
CLS3580
Corning® disposable spinner flasks
with vent caps, capacity 1 L, sterile
Sinonimo/i:
cell culture flasks, cell culture spinner flasks, scale up cell culture flasks
Scegli un formato
6 EA
1.480,00 €
1.480,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
6 EA
1.480,00 €
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
1.480,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
high-density polyethylene cap
polystyrene flask
vent cap
Sterilità
sterile
sterile
Caratteristiche
cap
closure type Vented cap
Confezionamento
case of 6
Produttore/marchio commerciale
Corning 3580
Capacità
1 L
Volume d’esercizio
300-1000 mL
Raccordi
cap for GL 45
GL 45 neck
Compatibilità
suitable for (scale-up cell culture; production of secreted proteins)
Descrizione generale
Convenience
The Corning disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
Confidence
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Compatibility
Made of ISO 10993 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Performance
The paddle size and height is optimized for each vessel size. A unique integrated magnet provides a smooth, even rotation at required speeds on any laboratory slow speed stirrer. Heat build-up in the vessel is reduced by means of a specially designed flange that raises the vessel off the stir-plate surface.
Conclusions
  • Disposable large volume spinner flasks are equivalent to conventional glass spinner flasks for cell growth and viability
  • A variety of cell types can be cultured in large volume disposable spinner flasks
  • Disposable spinner flasks maintain equal aeration as compared to standard glass spinner flasks
  • Cultured cells from glass and disposable spinner flasks are metabolically equivalent
  • Culured cells in disposable spinner flasks product equal amounts of secreted proteins as cells cultured in traditional glass spinner flasks
Corning disposable spinner flasks are designed to be single-use culture flask with no assembly required. These spinner flasks are suitable as scale-up cell culture vessels in addition to the production of secreted proteins.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)