Documenti fondamentali
BR780743
BRAND® screw cap for micro tubes
green
Sinonimo/i:
Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps, Tubes
Scegli un formato
1000 EA
123,00 €
123,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1000 EA
123,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
123,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
green
polypropylene
Confezionamento
pack of 1000 ea
Produttore/marchio commerciale
BRAND 780743
Descrizione generale
The BRAND® Screw Cap for Micro Tubes is designed specifically for use with micro tubes 780730 - 780734 and 780750 - 780754. Made from high-quality polypropylene (PP), these screw caps feature a silicone seal that ensures a secure closure, preventing leaks and contamination. They are suitable for a wide range of applications, withstanding temperatures from -196 °C to +121 °C. Certified as BIO-CERT® PCR QUALITY, these caps are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, making them ideal for sensitive molecular biology applications.
Note legali
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)