Documenti fondamentali
BR705220
BRAND® HandyStep® Touch and HandyStep® Touch S Charging Stand
Scegli un formato
1 EA
251,00 €
251,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
251,00 €
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.04
251,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Produttore/marchio commerciale
BRAND 705220
Descrizione generale
The charging stand for the HandyStep touch saves space on the laboratory bench and securely holds the device even with a PD-Tip II attached. It features inductive charging technology, which enables charging without cable clutter. Simply place the HandyStep touch into the stand, and the charging process begins.
The blue LED status indicator at the base of the charging stand shows the charging status:
LED lit: Device is charging
LED not lit: Device is fully charged
The charging stand can be used for both the HandyStep touch and the HandyStep touch S.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
HandyStep is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)