Documenti fondamentali
BR36954
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 2000 mL, neck joint: ST/NS 29/32, polypropylene stopper
Sinonimo/i:
BLAUBRAND®, USP, Volumetric flasks
Scegli un formato
1 EA
81,40 €
81,40 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
81,40 €
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
81,40 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
borosilicate glass
polypropylene stopper
Giunto
joint: ST/NS neck29/32
Caratteristiche
accuracy class A
Confezionamento
pack of 2
Produttore/marchio commerciale
BRAND 36954
Capacità
2000 mL
Descrizione generale
The BRAND® volumetric flask BLAUBRAND® USP conformity certified is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for a variety of laboratory applications. Each flask comes with a USP batch certificate, confirming its adherence to stringent quality and safety standards. This volumetric flask is an essential tool for precise analytical work, making it ideal for laboratories that require reliable and certified liquid handling solutions.
Caratteristiche e vantaggi
  • BLAUBRAND® 2000 mL volumetric flasks offer precise measurement of liquids with high accuracy even for partial volumes.
  • Computer-controlled systems guarantee the possible precision for fully automated production processes.
  • Individually calibrated for the best technical perfection.
  • USP volumetric instruments comply with the Class A error limits required by the United States Pharmacopeia.
  • The flask can be heated up to 250°C in a drying cabinet or sterilizer without any permanent volume changes.
  • The use of specially developed high-quality printing inks ensures a long service life.
  • The blue enamel used for BLAUBRAND® volumetric instruments offers an excellent color contrast. It represents the optimum combination of resistance and readability.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)