Documenti fondamentali
BAF199170002
Scienceware® vacuum aspirator bottle with trap
capacity 1.0 gal
Sinonimo/i:
bottle aspirator
Scegli un formato
1 EA
363,00 €
363,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
363,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.79
363,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F19917-0002
Capacità
1.0 gal
Descrizione generale
Make Quick Work of Large Jobs
For easy and continuous aseptic vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes. To use, simply insert a pipette tip, Pasteur pipette or serological pipette into the collection tube and connect vacuum line to vacuum source.
  • Chemical and corrosion resistant polypropylene bottle and epoxy coated steel frame
  • Will not crowd workspace, fume hood, or laminar flow hood
  • Quickly and conveniently aspirate and collect fluids using a vacuum source
  • Holds 3200 mL (108 oz) and the included trap prevents fluid flow into the vacuum source
  • Use to aspirate media, waste chemicals, cell washing solutions, and more
  • Ideal for use with dishes, plates, flasks and tubes
  • All parts, except the clear PVC tubing, are autoclavable at 121 °C (250 °F)
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)