Ugrás a tartalomra
Merck
  • Polymorphonuclear Neutrophil Functions are Differentially Altered in Amnestic Mild Cognitive Impairment and Mild Alzheimer's Disease Patients.
Polymorphonuclear Neutrophil Functions are Differentially Altered in Amnestic Mild Cognitive Impairment and Mild Alzheimer's Disease Patients.
Journal of Alzheimer's disease : JAD (2017-08-05)
Aurélie Le Page, Julie Lamoureux, Karine Bourgade, Eric H Frost, Graham Pawelec, Jacek M Witkowski, Anis Larbi, Gilles Dupuis, Tamàs Fülöp
KIVONAT
The mechanisms of neurodegeneration in Alzheimer's disease (AD) remain under investigation. Alterations in the blood-brain barrier facilitate exchange of inflammatory mediators and immune cells between the brain and the periphery in AD. Here, we report analysis of phenotype and functions of polymorphonuclear neutrophils (PMN) in peripheral blood from patients with amnestic mild cognitive impairment (aMCI, n = 13), patients with mild AD (mAD, n = 15), and healthy elderly controls (n = 13). Results showed an increased expression of CD177 in mAD but not in healthy or aMCI patients. IL-8 stimulated increased expression of the CD11b integrin in PMN of healthy subjects in vitro but PMN of aMCI and mAD patients failed to respond. CD14 and CD16 expression was lower in PMN of mAD but not in aMCI individuals relative to controls. Only PMN of aMCI subjects expressed lower levels of CD88. Phagocytosis toward opsonized E. coli was differentially impaired in PMN of aMCI and mAD subjects whereas the capacity to ingest Dextran particles was absent only in mAD subjects. Killing activity was severely impaired in aMCI and mAD subjects whereas free radical production was only impaired in mAD patients. Inflammatory cytokine (TNFα, IL-6, IL-1β, IL-12p70) and chemokine (MIP-1α, MIP-1β, IL-8) production in response to LPS stimulation was very low in aMCI and nearly absent in mAD subjects. TLR2 expression was low only in aMCI. Our data showed a differentially altered capacity of PMN of aMCI and mAD subjects to respond to pathological aggression that may impact impaired responses associated with AD development.