Ugrás a tartalomra
Merck
  • Inhibition of human esophageal squamous cell carcinomas by targeted silencing of tumor enhancer genes: an overview.
Inhibition of human esophageal squamous cell carcinomas by targeted silencing of tumor enhancer genes: an overview.
Cancer biology & medicine (2014-07-11)
Jalil Pirayesh Islamian, Mohsen Mohammadi, Behzad Baradaran
KIVONAT
Esophageal cancer has been reported as the ninth most common malignancy and ranks as the sixth most frequent cause of death worldwide. Esophageal cancer treatment involves surgery, chemotherapy, radiation therapy, or combination therapy. Novel strategies are needed to boost the oncologic outcome. Recent advances in the molecular biology of esophageal cancer have documented the role of genetic alterations in tumorigenesis. Oncogenes serve a pivotal function in tumorigenesis. Targeted therapies are directed at the unique molecular signature of cancer cells for enhanced efficacy with low toxicity. RNA interference (RNAi) technology is a powerful tool for silencing endogenous or exogenous genes in mammalian cells. Related results have shown that targeting oncogenes with siRNAs, specifically the mRNA, effectively reduces tumor cell proliferation and induces apoptotic cell death. This article will briefly review studies on silencing tumor enhancer genes related to the induction of esophageal cancer.