Ugrás a tartalomra
Merck
  • Post-transcriptional control of neurofilaments: New roles in development, regeneration and neurodegenerative disease.
Post-transcriptional control of neurofilaments: New roles in development, regeneration and neurodegenerative disease.
Trends in neurosciences (2009-11-13)
Ben G Szaro, Michael J Strong
KIVONAT
Neurofilament (NF) protein expression is coupled to axon development and the maintenance of neuronal homeostasis. Here, we present evidence that this tight regulation depends critically on post-transcriptionally regulated changes in NF mRNA transport, translation and stability. Recent studies have shown that post-transcriptional mechanisms modulate increases in NF gene transcription during axon regeneration to yield the final pattern of NF protein expression. Other recent work has found that post-transcriptional control of NFs shares elements with that of other axonal proteins and that its dysregulation contributes to amyotrophic lateral sclerosis. Such studies herald a novel approach to understanding how neurons coordinate the expressions of functionally related proteins and provide new insights into how the dysregulation of this control can lead to disease.