Ugrás a tartalomra
Merck
Systemic amyloidosis: lessons from β2-microglobulin.
The Journal of biological chemistry (2015-03-10)
Monica Stoppini, Vittorio Bellotti
KIVONAT
β2-Microglobulin is responsible for systemic amyloidosis affecting patients undergoing long-term hemodialysis. Its genetic variant D76N causes a very rare form of familial systemic amyloidosis. These two types of amyloidoses differ significantly in terms of the tissue localization of deposits and for major pathological features. Considering how the amyloidogenesis of the β2-microglobulin mechanism has been scrutinized in depth for the last three decades, the comparative analysis of molecular and pathological properties of wild type β2-microglobulin and of the D76N variant offers a unique opportunity to critically reconsider the current understanding of the relation between the protein's structural properties and its pathologic behavior.