Ugrás a tartalomra
Merck
Mechanism for nucleocytoplasmic shuttling of histone deacetylase 7.
The Journal of biological chemistry (2001-10-05)
H Y Kao, A Verdel, C C Tsai, C Simon, H Juguilon, S Khochbin
KIVONAT
Here we show that HDAC7, a member of the class II histone deacetylases, specifically targets several members of myocyte enhancer factors, MEF2A, -2C, and -2D, and inhibits their transcriptional activity. Furthermore, we demonstrate that DNA-bound MEF2C is capable of recruiting HDAC7, demonstrating that the HDAC7-dependent repression of transcription is not due to the inhibition of the MEF2 DNA binding activity. The data also suggest that the promoter bound MEF2 is potentially capable of remodeling adjacent nucleosomes via the recruitment of HDAC7. We have also observed a nucleocytoplasmic shuttling of HDAC7 and dissected the mechanism involved. In NIH3T3 cells, HDAC7 was primarily localized in the cytoplasm, essentially due to an active CRM1-dependent export of the protein from the nucleus. Interestingly, in HeLa cells, HDAC7 was predominantly nuclear. In these cells we could restore the cytoplasmic localization of HDAC7 by expressing CaMK I. This CaMK I-induced nuclear export of HDAC7 was abolished when three critical serines, Ser-178, Ser-344, and Ser-479, of HDAC7 were mutated. We show that these serines are involved in the direct interaction of HDAC7 with 14-3-3. Mutations of these serine residues weakened the association with 14-3-3 and dramatically enhanced the repression activity of HDAC7 in NIH3T3 cells, but not in HeLa cells. Data presented in this work clearly show that the signal dependent subcellular localization of HDAC7 is essential in controlling its activities. The data also show that the cellular concentration of factors such as 14-3-3, CaMK I, and other yet unknown molecules may determine the subcellular localization of an individual HDAC member in a cell type and HDAC-specific manner.