Ugrás a tartalomra
Merck
Glypican 3 overexpression in primary and metastatic Wilms tumors.
Virchows Archiv : an international journal of pathology (2014-11-05)
Maria Tretiakova, Debra L Zynger, Chunyan Luan, Nicole K Andeen, Laura S Finn, Masha Kocherginsky, Bin T Teh, Ximing J Yang
KIVONAT
Glypican 3 (GPC3), a heparan sulfate proteoglycan, plays a role in cell growth and differentiation. Mutations of the GPC3 gene are responsible for Simpson-Golabi-Behmel syndrome, which is characterized by anomalies of postnatal overgrowth and an increased risk of developing pediatric malignancies, mostly Wilms tumor and liver cancer. In order to understand the possible role of GPC3 in renal development and Wilms tumor formation, we analyzed messenger RNA (mRNA) and protein levels of GPC3 in sporadic Wilms tumors and compared it to normal kidneys and other common renal epithelial tumors. By using Affymetrix HGU133 oligonucleotide gene expression microarray data from 191 renal tumors and 12 normal kidneys, we found significant overexpression of GPC3 in Wilms tumors (p < 0.01), with 3.5-fold higher expression in comparison to normal kidneys and 6.5-fold higher than any type of renal tumors. The GPC3 gene product in Wilms tumor was further evaluated by immunohistochemistry and quantified by an automated image analysis. Cytoplasmic and membranous GPC3 immunoreactivity was present in 77 % of primary Wilms tumors (23/30), 93 % of metastatic Wilms tumors (13/14), 50 % of metanephric adenomas (4/8), 33 % of congenital mesoblastic nephromas (2/6), 100 % of nephrogenic rests (11/11), and 100 % of fetal kidneys (5/5). GPC3 staining was predominantly identified in blastemal and epithelial components of Wilms tumors, similar to that of fetal non-neoplastic kidney. All adult renal tumors (n = 60) and normal kidneys (n = 15) were GPC3 negative. These findings suggest the utility of GPC3 in differential diagnosis and follow-up of Wilms tumors. Our data also indicate that GPC3 is an oncofetal protein with a potential therapeutic value.