- Novel triptycene-based cylindrical macrotricyclic host: synthesis and complexation with paraquat derivatives.
Novel triptycene-based cylindrical macrotricyclic host: synthesis and complexation with paraquat derivatives.
Organic letters (2006-01-18)
Qian-Shou Zong, Chuan-Feng Chen
PMID16408877
KIVONAT
[reaction: see text] A novel triptycene-based cylindrical macrotricyclic polyether containing two dibenzo[24]crown-8 cavities has been synthesized and proved to be a highly efficient host for the complexation with paraquat derivatives. Consequently, a new kind of very stable pseudorotaxane-type complex was formed in solution and in the solid state.
ANYAGOK