Ugrás a tartalomra
Merck
  • 3,3'-Dihydroxyisorenieratene prevents UV-induced formation of reactive oxygen species and the release of protein-bound zinc ions in human skin fibroblasts.
3,3'-Dihydroxyisorenieratene prevents UV-induced formation of reactive oxygen species and the release of protein-bound zinc ions in human skin fibroblasts.
Molecular nutrition & food research (2009-10-29)
Kaya Lutter, Silke De Spirt, Sebastian Kock, Klaus-Dietrich Kröncke, Hans-Dieter Martin, Tanja Wagener, Wilhelm Stahl
KIVONAT
3,3'-Dihydroxyisorenieratene (DHIR) is a structurally unusual carotenoid exhibiting bifunctional antioxidant properties. It is synthesized by Brevibacterium linens, used in dairy industry for the production of red smear cheeses. The compound protects cellular structures against photo-oxidative damage and inhibits the UV-dependent formation of thymidine dimers. Here we show that DHIR prevents a UV-induced intracellular release of zinc ions from proteins in human dermal fibroblasts. The effect is correlated with a decreased formation of intracellular reactive oxygen species. In contrast, zinc release from cellular proteins induced by hyperthermia is not affected by pretreatment of cells with the antioxidant DHIR. It is suggested that the intracellular zinc release upon UV irradiation is due to oxidative modifications of the zinc ligands in proteins (e.g. cysteine) and that protection by DHIR is due to intracellular scavenging of reactive oxygen species generated in photo-oxidation.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Zinquin, ≥95% (HPLC), solid