Ugrás a tartalomra
Merck
  • The role of aligned polymer fiber-based constructs in the bridging of long peripheral nerve gaps.
The role of aligned polymer fiber-based constructs in the bridging of long peripheral nerve gaps.
Biomaterials (2008-05-02)
Young-Tae Kim, Valerie K Haftel, Satish Kumar, Ravi V Bellamkonda
KIVONAT
Peripheral nerve regeneration across long nerve gaps is clinically challenging. Autografts, the standard of therapy, are limited by availability and other complications. Here, using rigorous anatomical and functional measures, we report that aligned polymer fiber-based constructs present topographical cues that facilitate the regeneration of peripheral nerves across long nerve gaps. Significantly, aligned but not randomly oriented fibers elicit regeneration, establishing that topographical cues can influence endogenous nerve repair mechanisms in the absence of exogenous growth promoting proteins. Axons regenerated across a 17 mm nerve gap, reinnervated muscles, and reformed neuromuscular junctions. Electrophysiological and behavioral analyses revealed that aligned but not randomly oriented constructs facilitated both sensory and motor nerve regeneration, significantly improved functional outcomes. Additionally, a quantitative comparison of DRG outgrowth in vitro and nerve regeneration in vivo on aligned and randomly oriented fiber films clearly demonstrated the significant role of sub-micron scale topographical cues in stimulating endogenous nerve repair mechanisms.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
α-Bungarotoxin-tetramethylrhodamine from Bungarus multicinctus (Formosan Banded Krait)