Ugrás a tartalomra
Merck
  • Synthesis of Strongly Fluorescent Imidazole Derivatives: Structure Property Studies, Halochromism and Fluorescent Photoswitching.
Synthesis of Strongly Fluorescent Imidazole Derivatives: Structure Property Studies, Halochromism and Fluorescent Photoswitching.
Journal of fluorescence (2019-11-16)
Anu Kundu, Subramanian Karthikeyan, Dohyun Moon, Savarimuthu Philip Anthony
KIVONAT
New series of methoxy and hydroxyl group substituted triphenylamine (TPA)-imidazole fluorescent molecules (5-(diphenylamino)-2-(1H-phenanthro[9,10-d]imidazol-2-yl)phenol (1), 5-(diphenylamino)-2-(1-phenyl-1H-phenanthro[9,10-d]imidazol-2-yl)phenol (2), 5-(diphenylamino)-2-(4,5-diphenyl-1H-imidazol-2-yl)phenol (3), 5-(diphenylamino)-2-(1,4,5-triphenyl-1H-imidazol-2-yl)phenol (4), N-(3-methoxy-4-(1H-phenanthro[9,10-d]imidazol-2-yl)phenyl)-N-phenylbenzenamine (5), N-(3-methoxy-4-(1-phenyl-1H-phenanthro[9,10-d]imidazol-2-yl)phenyl)-N-phenylbenzene amine (6), and N-(3-methoxy-4-(4,5-diphenyl-1H-imidazol-2-yl)phenyl)-N-phenylbenzenamine (7)) have been synthesized that exhibited strong solution fluorescence and molecular structure and conformation controlled fluorescence photoswitching, solid state fluorescence and halochromism. Hydroxyl substituted molecules (1-4) showed moderate to strong fluorescence in solution depend on solvent polarity and very weak solid state fluorescence. Methoxy substituted molecules (5-7) displayed strong fluorescence both in solution and solid state. Solid state structural studies revealed strong intramolecular H-bonding in the crystal lattice. Interestingly, highly twisted structure (6) showed rare light induced reversible fluorescence switching in CHCl3. The observation of isobestic point in time dependent fluorescence photoswitching studies indicated structural isomer conversion. Further, acid sensitive imidazole nitrogen has been made use to demonstrate solid state fluorescence switching via halochromism. Thus the present studies attempted to develop new fluorescent molecules and establish structure-property relationship for designing fluorescence switching materials. Graphical Abstract Molecular structure controlled solid state fluorescence, halochromism and a rare fluorescence photoswitching in chloroform have been observed with triphenylamine-imidazole derivatives.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
4-(Diphenylamino)benzaldehyde, 97%