Ugrás a tartalomra
Merck
  • Circulating Cell-Free mtDNA Contributes to AIM2 Inflammasome-Mediated Chronic Inflammation in Patients with Type 2 Diabetes.
Circulating Cell-Free mtDNA Contributes to AIM2 Inflammasome-Mediated Chronic Inflammation in Patients with Type 2 Diabetes.
Cells (2019-04-11)
Jung Hwan Bae, Seung Ii Jo, Seong Jin Kim, Jong Min Lee, Ji Hun Jeong, Jeong Suk Kang, Nam-Jun Cho, Sang Soo Kim, Eun Young Lee, Jong-Seok Moon
KIVONAT
Mitochondrial dysfunction has been implicated in the pathogenesis of insulin resistance and type 2 diabetes. Damaged mitochondria DNA (mtDNA) may have a role in regulating hyperglycemia during type 2 diabetes. Circulating cell-free mitochondria DNA (ccf-mtDNA) was found in serum and plasma from patients and has been linked to the prognosis factors in various human diseases. However, the role of ccf-mtDNA in chronic inflammation in type 2 diabetes is unclear. In this study, we hypothesized that the ccf-mtDNA levels are associated with chronic inflammation in patients with type 2 diabetes. The mtDNA levels were elevated in the plasma from patients with type 2 diabetes compared to healthy subjects. The elevated mtDNA levels were associated with interleukin-1 (IL-1)β levels in patients with type 2 diabetes. The mtDNA, from patients with type 2 diabetes, induced absent in melanoma 2 (AIM2) inflammasome-dependent caspase-1 activation and IL-1β and IL-18 secretion in macrophages. Our results suggest that the ccf-mtDNA might contribute to AIM2 inflammasome-mediated chronic inflammation in type 2 diabetes.