Ugrás a tartalomra
Merck
  • Effects of L-carnitine on morphology and cellular parameters of hen erythrocytes.
Effects of L-carnitine on morphology and cellular parameters of hen erythrocytes.
Polish journal of veterinary sciences (2019-01-04)
K Kasperek, A Bownik, S Knaga, A Szabelak, B Ślaska, M Kwiecień, G Jeżewska-Witkowska
KIVONAT
The aim of our study was to determine the influence of L-carnitine (L-CAR) on the cellular parameters of hen erythrocytes during a 48 hour exposure to L-CAR at concentrations of 25, 50 and 100 µg/mL in nutrient-deficient medium. Cell morphology, haemolysis, caspase 3/7 activity and glucose uptake (GU) were determined. The results showed a lower percentage of apoptotic cells and decreased haemolysis of erythrocytes treated for 48 hours at all the concentrations of L-CAR. The amino acid at 50 µg/mL inhibited the activity of proapoptotic caspase 3/7; however, it increased GU. In contrast, caspase 3/7 level was increased but GU was decreased in erythrocytes treated with 100 µg/mL of L-CAR when compared to the control. It may be hypothesized that reduction of apoptotic changes in hen erythrocytes may result from increased GU.