Ugrás a tartalomra
Merck
  • Inhibition of oncogenic cap-dependent translation by 4EGI-1 reduces growth, enhances chemosensitivity and alters genome-wide translation in non-small cell lung cancer.
Inhibition of oncogenic cap-dependent translation by 4EGI-1 reduces growth, enhances chemosensitivity and alters genome-wide translation in non-small cell lung cancer.
Cancer gene therapy (2018-11-14)
Arpita De, Blake A Jacobson, Mark S Peterson, Margaret E Stelzner, Joe Jay-Dixon, Marian G Kratzke, Manish R Patel, Peter B Bitterman, Robert A Kratzke
KIVONAT
Hyperactivation of eIF4F-mediated translation occurs in many if not all cancers. As a consequence, cancer cells aberrantly enhance expression of malignancy-related proteins that are involved in cell cycle progression, angiogenesis, growth, and proliferation. With this in mind eIF4F is a promising molecular target for therapeutics that counteract pathological eIF4F activity. Here we used 4EGI-1, a small-molecule inhibitor of cap-mediated translation that disrupts formation of the eukaryotic initiation factor 4F (eIF4F) complex to treat non-small cell lung cancer (NSCLC). Treatment of cells with 4EGI-1 reduced cell proliferation, decreased cap-dependent complex formation, induced apoptosis, enhanced sensitivity to gemcitabine, and altered global cellular translation. Suppression of cap-dependent translation by 4EGI-1 resulted in diminished expression of oncogenic proteins c-Myc, Bcl-2, cyclin D1, and survivin, whereas β-actin expression was left unchanged. In light of these results, small-molecule inhibitors like 4EGI-1 alone or with chemotherapy should be further evaluated in the treatment of NSCLC.