Ugrás a tartalomra
Merck
  • Efficient Charge Carrier Separation in l-Alanine Acids Derived N-TiO2 Nanospheres: The Role of Oxygen Vacancies in Tetrahedral Ti4+ Sites.
Efficient Charge Carrier Separation in l-Alanine Acids Derived N-TiO2 Nanospheres: The Role of Oxygen Vacancies in Tetrahedral Ti4+ Sites.
Nanomaterials (Basel, Switzerland) (2019-05-08)
Yongjuan Chen, Xiu Luo, Yao Luo, Peiwen Xu, Jiao He, Liang Jiang, Junjie Li, Zhiying Yan, Jiaqiang Wang
KIVONAT
N-doped TiO2 with oxygen vacancies exhibits many advantages for photocatalysis, such as enhanced visible light absorbency, inhibition of the photogenerated charge carrier recombination, etc. However, preparation of N-doped TiO2 with oxygen vacancies under mild conditions is still a challenge. Herein, N-doped TiO2 nanospheres with tetrahedral Ti4+ sites were synthesized by using dodecylamine as template and assisted by l-alanine acids. The obtained samples were characterized by X-ray powder diffraction (XRD), scanning electron microscopy (SEM), transmission electron microscopy (TEM), and UV-Vis diffuse reflectance spectra (UV-Vis DRS). It was found that the dodecylamine as a neutral surfactant controlled the structure of TiO2 spherical, while l-alanine acids provided a nitrogen source. The existence of tetrahedral Ti4+ sites in N-doped TiO2 was also confirmed. The N-doped TiO2 sample with tetrahedral Ti4+ sites exhibited significantly improved photocatalytic performance for degradation of methylene blue solution under UV light or visible light irradiation. A combined time-resolved infrared (IR) spectroscopy study reveals that the enhanced photocatalytic performance could be attributed to a large amount of photogenerated charge carriers and efficient charge separation. It is demonstrated that the shallow donor state produced by oxygen vacancies of tetrahedral Ti4+ sites can effectively promote separation of charge carriers besides capturing electrons.