Ugrás a tartalomra
Merck
  • RNAi-mediated silencing of VEGF-C inhibits non-small cell lung cancer progression by simultaneously down-regulating the CXCR4, CCR7, VEGFR-2 and VEGFR-3-dependent axes-induced ERK, p38 and AKT signalling pathways.
RNAi-mediated silencing of VEGF-C inhibits non-small cell lung cancer progression by simultaneously down-regulating the CXCR4, CCR7, VEGFR-2 and VEGFR-3-dependent axes-induced ERK, p38 and AKT signalling pathways.
European journal of cancer (Oxford, England : 1990) (2011-06-18)
Yukuan Feng, Jing Hu, Jing Ma, Kejian Feng, Xiaoli Zhang, Shucai Yang, Wei Wang, Jianguo Zhang, Yafang Zhang
KIVONAT
Vascular endothelial growth factor C (VEGF-C) expression is associated with the malignant tumour phenotype making it an attractive therapeutic target. We investigated the biological roles of VEGF-C in tumour growth, migration, invasion and explored the possibility of VEGF-C as a potential therapeutic target for the treatment of non-small cell lung cancer (NSCLC). A lentivirus-mediated RNA interference (RNAi) technology was used to specifically knockdown the expression of VEGF-C in A549 cells. Quantitative reverse transcriptase-polymerase chain reaction, flow cytometry, Western blot, immunohistochemistry, cellular growth, migration, invasion and ELISA assays were used to characterise VEGF-C expression in vitro. A lung cancer xenograft model in nude mice was established to investigate whether knockdown of VEGF-C reduced tumour growth in vivo. Silencing of VEGF-C suppressed tumour cell growth, migration and invasion in vitro; suppressed tumour growth, angiogenesis and lymphangiogenesis by tail vein injection of lentivirus encoded shRNA against VEGF-C in vivo. More importantly, silencing of VEGF-C also trapped the VEGFR-2, VEGFR-3, CXCR4, CCR7-dependent axes, and down-regulated the AKT, ERK and p38 signalling pathways. These results suggest that VEGF-C has a multifaceted role in NSCLC growth, migration and invasion; that VEGF-C-mediated autocrine loops with their cognate receptors and chemokine receptors are significant factors affecting tumour progression; and that RNAi-mediated silencing of VEGF-C represents a powerful therapeutic approach for controlling NSCLC growth and metastasis.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), fluorimetric, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with fluorescent detection.
Sigma-Aldrich
QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), fluorimetric, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.