Ugrás a tartalomra
Merck
  • Vertical Alignment of Liquid Crystals Over a Functionalized Flexible Substrate.
Vertical Alignment of Liquid Crystals Over a Functionalized Flexible Substrate.
Scientific reports (2018-06-13)
B Sivaranjini, R Mangaiyarkarasi, V Ganesh, S Umadevi
KIVONAT
A simple and effective approach for vertical alignment of liquid crystals (LCs) over a functionalized transparent flexible substrate is described. Surface characterization of this commercially available plastic substrate through X-ray photoelectron spectroscopy (XPS) and attenuated total reflection infrared spectroscopy (ATR-IR) indicated that cellulose acetate is main component of the transparent substrate. This substrate was chemically functionalized with a suitable LC compound. A trimethoxysilane terminated new rod-shaped mesogen is synthesized and covalently attached to the pre-treated film through silane condensation reaction. LC functionalization of the polymer film is confirmed through contact angle (CA), atomic force microscopy (AFM), XPS and ATR-IR spectroscopy studies. Versatility of the LC modified flexible substrates for the alignment of bulk LC sample at substrate-LC interface was assessed for nematic (N) and smectic A (SmA) phases. Remarkably, LC functionalized cellulose acetate films were found to be highly efficient in assisting a perfect homeotropic alignment of LCs (for both, a room temperature N and a high temperature SmA phase) over the entire area of the LC sample under observation indicating their superior aligning ability in comparison to their unmodified and octadecyltrimethoxysilane (OTS) modified counterparts. The demonstrated method of surface modification of flexible polymer film is easy, surface modified substrates are stable for several months, retained their aligning ability intact and more importantly they are reusable with maximum delivery.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
1,3-Divinyltetramethyldisiloxane, 97%