Skip to Content
Merck
All Photos(1)

Key Documents

BGM Cell Line

90092601, African green monkey kidney, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

BGM Cell Line, 90092601, from African green monkey kidney, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

African green monkey kidney

packaging

tube of 5 μg 90092601-DNA-5UG
pkg of vial of cells 90092601-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Monkey African green kidney

Cell Line Description

Used for the isolation of water borne viruses e.g. polio 1, 2, 3, Echo 3, 6, 7, 9, 12, 27, Coxsackie A9 + B1, 2, 3 and Reo 1.

Application

Virus studies: polio, Echo, Coxsackie and Reo viruses

Culture Medium

EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 10% FBS / FCS (F2442).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 to 1:20, i.e., seeding at 5x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service