跳转至内容
Merck
  • Green chemoselective synthesis of thiazolo[3,2-a]pyridine derivatives and evaluation of their antioxidant and cytotoxic activities.
Green chemoselective synthesis of thiazolo[3,2-a]pyridine derivatives and evaluation of their antioxidant and cytotoxic activities.
Bioorganic & medicinal chemistry letters (2009-09-05)
Feng Shi, Chunmei Li, Ming Xia, Kangjie Miao, Yanxia Zhao, Shujiang Tu, Weifa Zheng, Ge Zhang, Ning Ma
摘要
The green chemoselective synthesis of thiazolo[3,2-a]pyridine derivatives was achieved in water via microwave-assisted three-component reactions of malononitrile, aromatic aldehydes and 2-mercaptoacetic acid with molar ratios of 2:1:1.5 and 2:2.2:1, respectively. These compounds were subject to the experiments of antioxidant activity and cytotoxicity to carcinoma HCT-116 cells and mice lymphocytes. Nearly all of the tested compounds possessed potent capacities for scavenging free radicals. In addition, most of these compounds showed cytotoxicity to HCT-116 cells and mice lymphocytes with no selectivity. Of these, only thiazolo[3,2-a]pyridine derivative 5d suggested selective cytotoxicity to tumor cell line HCT-116 cells.
材料
货号
品牌
产品描述
Sigma-Aldrich
L -抗坏血酸, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L -抗坏血酸, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L -抗坏血酸, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L -抗坏血酸, reagent grade, crystalline
Supelco
L -抗坏血酸, analytical standard
Sigma-Aldrich
L -抗坏血酸, reagent grade
Sigma-Aldrich
L -抗坏血酸, meets USP testing specifications
Sigma-Aldrich
L -抗坏血酸, FCC, FG
Sigma-Aldrich
L -抗坏血酸, 99%
Sigma-Aldrich
L -抗坏血酸, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L -抗坏血酸, ACS reagent, ≥99%
Sigma-Aldrich
L -抗坏血酸, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L -抗坏血酸, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L -抗坏血酸, tested according to Ph. Eur.