28527-U
1 of 10
Supelco
27212
预组装螺纹口样品瓶,带聚丙烯顶部开口瓶盖
USP
1601102
Residual Solvents Mixture - Class 1
28522-U
ATIS 吹扫和捕集/增湿器模块
28520-U
ATIS 吸附管进样器系统 110V
28523-U
带微型连接头的 ATIS 萃取玻璃器具
Z777202
Proteomix® SAX-NP10 Strong Anion Exchange Guard Cartridge and Holder
PHR1063
溶剂残留混标 I
28526-U
ATIS 标准进样玻璃器具备件
1.51912
Purospher® STAR Si (5 µm)
58365
SUPELCOSIL™ LC-Si HPLC 柱
air monitoringenvironmentalindustrial hygiene
空气监测装置和采样管
Choose from one of the most recent versions:
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 客户支持