Merck
25806
Supelco
T 形气体管路过滤器
brass, 1/4 in. male
登录查看公司和协议定价
About This Item
UNSPSC代码:
41112500
物料
brass
配件
1/4 in. male
正在寻找类似产品? 访问 产品对比指南
一般描述
This gas line filter contains a replaceable 7 μm sintered stainless steel frit, and will remove dust and small particles that could otherwise foul downstream needle valves. Nuts and ferrules included.
历史批次信息供参考:
分析证书(COA)