主要文件
GE28-9232-44
MidiSpin Adapter
Cytiva 28-9232-44, pack of 10 pieces
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115700
NACRES:
NA.56
特点
wetted part: no
包装
pack of 10 pieces
制造商/商品名称
Cytiva 28-9232-44
一般描述
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
特点和优势
  • Convenient adapters, enable spinning of all gravity columns.
储存分类代码
13 - Non Combustible Solids
历史批次信息供参考:
分析证书(COA)
访问文档库
实验方案
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.