主要文件
Z158372
Jacketed still body, with Vigreux indents
fits Ace jacketed short path distillation apparatus
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104812
NACRES:
NB.43
制造商/商品名称
Ace Glass 931752
一般描述
Replacement part for Ace jacketed short-path distillation apparatus. Vigreux indents run the entire length of the column and act as the packing for the column.
历史批次信息供参考:
分析证书(COA)