主要文件
Z108340
Leather gloves
size L
登录查看公司和协议定价
About This Item
UNSPSC代码:
46181504
NACRES:
NB.35
物料
brown gloves
leather
尺寸
L
无菌性
non-sterile
特点
latex free
powder-free
reusable
包装
pack of 1 pair
制造商/商品名称
Sigma-Aldrich
正在寻找类似产品? 访问 产品对比指南
一般描述
Constructed of specially treated leather that stays supple even at temperature extremes of -70 to 150°C. Remarkably durable, moisture, heat, and flame resistant. Provided with an elastic cinch for a no-slip fit.
历史批次信息供参考:
分析证书(COA)