主要文件
MT51340009
Mettler Toledo DX series ISE half-cell membrane kit
For Calcium ISE
登录查看公司和协议定价
About This Item
UNSPSC代码:
39121436
NACRES:
NB.79
制造商/商品名称
Mettler-Toledo 51340009
正在寻找类似产品? 访问 产品对比指南
一般描述
DX series ion specific membrane kit. Mount the new membrane onto the shaft of your ISE half-cell, and you have a new ISE!
Kit contains one membrane plus electrolyte
历史批次信息供参考:
分析证书(COA)