CLS2157100TJ
别名:
centrifuge tubes
1 of 10
WHAWB120206
QIAcard™ Indicating FTA™ Cards
Supelco
214477
硅胶
CLS30221L
Pyrex® 刻度量筒,单公制,白色搪瓷刻度
WHAWB120205
QIAcard™ Non-Indicating FTA™ Cards
64825
Soxhlet 萃取设备
CLS29821L
带 Pyrex® 塞子的 Pyrex® 刻度量筒
BR362138
BRAND® BLAUBRAND® ASTM 100 mL离心管
Millipore
SLHV033N
Millex™ PVDF 针式过滤器
24633
带有Thermogreen® LB-2胶垫的1L Tedlar® PLV气体采样袋
BR362338
Corning 2157-100TJ
100 mL
样品处理
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.