主要文件
BR23868
BRAND®备用滴定管延长管,适用于紧凑型自动滴定管
50 mL, SILBERBRAND
别名:
Accessories, Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121808
NACRES:
NB.43
物料
amber AR-Glass
包装
pack of 1 ea
制造商/商品名称
BRAND 23868
测量范围
50 mL
滴管长度
685 mm
体积
50 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
The BRAND® spare burette length for the compact automatic burette is made from high-quality Boro 3.3 clear glass, ensuring durability and clarity for precise measurements. Designed specifically for compatibility with BLAUBRAND® and SILBERBRAND compact automatic burettes, this spare part maintains the performance standards required for accurate liquid dispensing.
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)