EP5495500006
Rotor for Eppendorf® 5425 Centrifuge
FA-24x2, lid
About This Item
Productos recomendados
feature
lid
FA-24x2
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5495500006
parameter
15,060 max. RCF (21,300 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5425
General description
Rotor for Eppendorf(R) 5425 Centrifuge
Features and Benefits
- Aerosol tight rotor FA-24x2 for vessels, 24 x 1.5/2.0 ml
- Features aerosol-tight rotor lid
- Compatible with centrifuge 5425/R
- Autoclavable at 121 °C for 20 min
Legal Information
Eppendorf is a registered trademark of Eppendorf AG