EP5495400001
Eppendorf® FA-24x2 rotor
for 24 x 1.5/2.0-mL tubes incl. rotor cover, aerosol-tight, Centrifuge 5420
About This Item
Productos recomendados
feature
for 24 x 1.5/2.0-mL tubes incl. rotor cover, aerosol-tight, Centrifuge 5420
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 5495400001
parameter
10,000 rpm speed
storage temp.
room temp
Legal Information
Eppendorf is a registered trademark of Eppendorf AG