Merck
GBL204064
Grace Bio-Labs FlexWell removable incubation chambers
L × W × thickness 3.5 mm × 3.5 mm × 1.8 mm, black silicone, adhesive one side, wells 64
Synonym(e):
flexible silicone slide gasket, silicone multiwell gasket
Größe auswählen
5 EA
107,00 €
107,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
5 EA
107,00 €
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
107,00 €
Check Cart for Availability
Materialien
black silicone
Sterilität
non-sterile
Verpackung
pack of 5 ea
Hersteller/Markenname
Grace Bio-Labs 204064
L × B × Dicke
3.5 mm × 3.5 mm × 1.8 mm
Wellbereich
0.12 cm2
Wells
64
Allgemeine Beschreibung
FlexWell incubation chambers provide a convenient way to conduct binding reactions on protein microarrays using nitrocellulose coated ONCYTE® Film slides.
  • Silicone FlexWell forms wells on slides using a clean release adhesive to isolate up to 16 specimens per slide
  • Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SMS compliant microtiter plate
  • Gaskets may be sealed for mixing and to prevent evaporation
Rechtliche Hinweise
FlexWell is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)