Merck
F7016
Sigma-Aldrich
Flumequin
≥97.0% (HPLC)
Synonym(e):
9-Fluoro-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic acid
About This Item
Empirische Formel (Hill-System):
C14H12FNO3
CAS-Nummer:
Molekulargewicht:
261.25
Beilstein:
490724
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
51102829
PubChem Substanz-ID:
NACRES:
NA.85
Assay
≥97.0% (HPLC)
Form
powder
Farbe
white to off-white
Wirkungsspektrum von Antibiotika
Gram-negative bacteria
Gram-positive bacteria
Wirkungsweise
DNA synthesis | interferes
enzyme | inhibits
SMILES String
CC1CCc2cc(F)cc3C(=O)C(=CN1c23)C(O)=O
InChI
1S/C14H12FNO3/c1-7-2-3-8-4-9(15)5-10-12(8)16(7)6-11(13(10)17)14(18)19/h4-7H,2-3H2,1H3,(H,18,19)
InChIKey
DPSPPJIUMHPXMA-UHFFFAOYSA-N
Allgemeine Beschreibung
Chemical structure: fluoroquinolone
Biochem./physiol. Wirkung
Flumequine inhibits topoisomerases, which are needed for the transcription and replication of bacterial DNA. The inhibition of the topoisomerases results in strand breakage of the bacterial chromosome, supercoiling, and resealing. Therefore, DNA replication and transcription is inhibited .
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
Eyeshields, Gloves, type N95 (US)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Etienne Giraud et al.
Journal of medical microbiology, 53(Pt 9), 895-901 (2004-08-18)
The mechanisms of resistance to quinolone and epidemiological relationships among A. salmonicida strains isolated from diseased fish in French marine farms from 1998 to 2000 were investigated. The quinolone resistance-determining regions of the gyrA and parC genes of 12 clinical
Yoko Kashida et al.
Toxicological sciences : an official journal of the Society of Toxicology, 69(2), 317-321 (2002-10-16)
In order to elucidate the tumor-initiating potential of flumequine (FL) in the liver, male C3H mice were given dietary administration of 4000 ppm FL throughout the study or for 2 weeks at the initiation stage, and then received 2 intraperitoneal
I Guillemin et al.
Microbiology (Reading, England), 145 ( Pt 9), 2527-2532 (1999-10-12)
The DNA gyrases from Mycobacterium avium, Mycobacterium smegmatis and Mycobacterium fortuitum bv. peregrinum, which are species naturally resistant, moderately susceptible and susceptible to fluoroquinolones, respectively, were purified by affinity chromatography on novobiocin-Sepharose columns. The DNA gyrase inhibiting activities (IC50 values)
E Giraud et al.
Antimicrobial agents and chemotherapy, 43(9), 2131-2137 (1999-09-03)
The occurrence of mutations in the genes coding for gyrase (gyrA and gyrB) and topoisomerase IV (parE and parC) of Salmonella typhimurium experimental mutants selected in vitro and in vivo and of 138 nalidixic acid-resistant Salmonella field isolates was investigated.
C Ferraresi et al.
Poultry science, 92(12), 3158-3165 (2013-11-16)
Flumequine (FLU) is used in the treatment of systemic bacterial infections in poultry, including colibacillosis, which is a common disease in turkeys. The pharmacokinetic (PK) behavior of FLU administered to 32 healthy turkeys as an oral bolus via gavage or