Merck
Z678309
Finnpipette® F2 variable volume pipette
volume 2-20 μL , Micro-tip
Größe auswählen
1 EA
384,00 €
384,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
384,00 €
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
384,00 €
Check Cart for Availability
Leistungsmerkmale
color-coded turquoise
electronic: no
single-channel
Verpackung
pack of 1 ea
Hersteller/Markenname
Finn 4642050
Volumen
2-20 μL , Micro-tip
Allgemeine Beschreibung
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50μl and below
Rechtliche Hinweise
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)