Merck
Z136255
Tubing sizer
determines I.D. and O.D. of glass tubing, test tubes, etc
About This Item
UNSPSC-Code:
41105108
NACRES:
NB.35
Materialien
polypropylene
Hersteller/Markenname
Bel-Art
Messbereich
1.5-16 mm
L × B × H
120 mm × 50 mm × 12 mm
Allgemeine Beschreibung
Determines I.D. and O.D. of glass tubing, test tubes, etc. For I.D., select the closest pin size; for O.D., choose closest well size.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)