Merck
SLW1060/04D
Pyrex® Watch glasses
nominal diameter 40 mm
Synonym(e):
Pyrex, SciLabware 1060/04D
Größe auswählen
10 EA
176,00 €
176,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
176,00 €
About This Item
UNSPSC-Code:
41121801
NACRES:
NB.43
176,00 €
Check Cart for Availability
Verpackung
pkg of 10
Hersteller/Markenname
1060/04D
SciLabware
Nenndurchmesser
40 mm
Allgemeine Beschreibung
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Leistungsmerkmale und Vorteile
  • Complies with DIN 12341
  • Manufactured from borosilicate glass 3.3
  • Heat resistant
  • Fire-polished edges to reduce chipping and breakage
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)