Merck
BR90228
BRAND® ETFE beaker with spout, low form
volume 50 mL, with graduation
Synonym(e):
Beakers, Plastics
Größe auswählen
1 EA
15,50 €
15,50 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
15,50 €
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
15,50 €
Check Cart for Availability
Materialien
ETFE
Leistungsmerkmale
with graduation
Hersteller/Markenname
BRAND 90228
Volumen
50 mL
Allgemeine Beschreibung
Made from high-quality ETFE (ethylene-tetrafluoroethylene), this translucent beaker offers excellent chemical resistance, making it ideal for handling various substances in laboratory settings. The low form provides stability and ease of use, allowing for efficient mixing and pouring of liquids. Clearly marked graduations facilitate accurate measurement of liquids, ensuring precision in experiments and formulations. The beaded rim enhances strength and provides a comfortable grip, making handling safer and more efficient.
Sonstige Hinweise
Browse all plastic beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)