Merck
BR23846
BRAND® spare burette length for compact automatic burette
25 mL, BLAUBRAND®
Synonym(e):
Accessories, Automatic Burettes, BLAUBRAND®, Burettes, Compact Automatic Burettes
Größe auswählen
1 EA
52,90 €
52,90 €
Versandbereit am22. April 2025Details
Größe auswählen
Ansicht ändern
1 EA
52,90 €
About This Item
UNSPSC-Code:
41121808
NACRES:
NB.43
52,90 €
Versandbereit am22. April 2025Details
Materialien
clear borosilicate glass 3.3
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 23846
Messbereich
25 mL
Bürette L
690 mm
Volumen
25 mL
Allgemeine Beschreibung
The BRAND® spare burette length for the compact automatic burette is made from high-quality Boro 3.3 clear glass, ensuring durability and clarity for precise measurements. Designed specifically for compatibility with BLAUBRAND® and SILBERBRAND compact automatic burettes, this spare part maintains the performance standards required for accurate liquid dispensing.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)