Merck
BR13943
BRAND® spare burette tubes for compact burette
size 10 mL, BLAUBRAND®, clear borosilicate glass 3.3
Synonym(e):
Accessories, BLAUBRAND®, Burettes, Compact Burettes
Größe auswählen
1 EA
30,60 €
30,60 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
30,60 €
About This Item
UNSPSC-Code:
41121808
NACRES:
NB.43
30,60 €
Check Cart for Availability
Materialien
clear borosilicate glass 3.3
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 13943
Länge
700 mm
Größe
10 mL
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 clear glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)