Skip to Content
Merck

Skip To

SML0908

CCG-50014

≥98% (HPLC)

Synonym(s):

4-[(4-Fluorophenyl)methyl]-2-(4-methylphenyl)-1,2,4-thiadiazolidine-3,5-dione

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C16H13FN2O2S
CAS Number:
Molecular Weight:
316.35
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality level:


Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C[n]2[c]([s][n]([c]2=O)c3ccc(cc3)C)=O

InChI

1S/C16H13FN2O2S/c1-11-2-8-14(9-3-11)19-15(20)18(16(21)22-19)10-12-4-6-13(17)7-5-12/h2-9H,10H2,1H3

InChI key

QUIIIYITNGOFEI-UHFFFAOYSA-N

Biochem/physiol Actions

CCG-50014 is an inhibitor of Regulators of G protein Signaling (RGS) proteins RGS4 and RGS8, which bind to the Gα -subunits of activated heterotrimeric G-proteins, and accelerate the rate of GTP hydrolysis, acting as GTPase activating proteins. CCG-50014 irreversibly binds and destabilizes RGS proteins with no affect on Gα0, while blocking GTPase activity and Gα0-dependent translocation of RGS4 to the membrane.
CCG-50014 is an inhibitor of Regulators of G protein Signaling (RGS) proteins RGS4 and RGS8.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1207SML1784SML0994
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 10 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.





Global Trade Item Number

SKUGTIN
SML0908-25MG04061832982823
SML0908-5MG04061832982830

Questions

Reviews

No rating value

Active Filters