Skip to Content
Merck

Skip To

SML0994

DFU

≥98% (HPLC)

Synonym(s):

3-(3-Fluorophenyl)-5,5-dimethyl-4-[4-(methylsulfonyl)phenyl]-2(5H)-furanone, 5,5-Dimethyl-3-(3 fluorophenyl)-4-(4 methylsulphonal) phenyl-2 (5H)-furanone, CID 9906892

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
CZK 8,220.00

About This Item

Empirical Formula (Hill Notation):
C19H17FO4S
CAS Number:
Molecular Weight:
360.40
NACRES:
NA.77
UNSPSC Code:
12352100
Assay:
≥98% (HPLC)
Form:
powder

CZK 8,220.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

DFU is an orally available highly selective cyclo-oxygenase-2 (COX-2) inhibitor that potently inhibits lipopolysaccharide (LPS)-induced PGE2 production in a human whole blood assay.
DFU is an orally available highly selective cyclo-oxygenase-2 (COX-2) inhibitor.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1355SML1990SML2887
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

yellow to orange

color

white to beige

color

white to beige


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters