Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
human heart (ventricles)
packaging
pkg of 500,000 cells
morphology
( endothelial)
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
diabetes; hypertension; ischemia/reperfusion injury
shipped in
ambient
storage temp.
room temp
General description
Application
Biochem/physiol Actions
Preparation Note
Analysis Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| relevant disease(s) diabetes; hypertension; ischemia/reperfusion injury | relevant disease(s) diabetes; hypertension; ischemia/reperfusion injury | relevant disease(s) diabetes; hypertension; ischemia/reperfusion injury | relevant disease(s) - |
| biological source human heart (ventricles) | biological source human heart (ventricles) | biological source human heart (ventricles) | biological source human coronary artery |
| morphology ( endothelial) | morphology ( endothelial) | morphology (endothelial) | morphology ( endothelial) |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) - | technique(s) cell culture | mammalian: suitable |
| shipped in ambient | shipped in dry ice | shipped in dry ice | shipped in ambient |
| packaging pkg of 500,000 cells | packaging pkg of 500,000 cells | packaging pkg of 1,000,000 cells | packaging pkg of 500,000 cells |
Still not finding the right product?
Explore all of our products under Human Cardiac Microvascular Endothelial Cells (HCMEC)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Protokol buněčné kultivace: test migrace a invaze endoteliálních buněk přes stěnu (transwell) používaný ke studiu angiogeneze a metastazování nádorových buněk. Prozkoumejte více než 350 produktů PromoCell.
Cell culture protocol: the endothelial cell transwell migration and invasion assay used to study angiogenesis and cancer cell metastasis. Explore over 350 PromoCell products.
Related Content
Protokol buněčné kultivace: test tvorby endoteliálních trubic ke studiu angiogeneze pomocí HUVEC a dalších typů endoteliálních buněk. Prozkoumejte více než 350 produktů PromoCell.
Cell culture protocol: the endothelial tube formation assay to study angiogenesis using HUVECs and other endothelial cell types. Explore over 350 PromoCell products.

