Skip to Content
Merck

Skip To

C-12222

Human Coronary Artery Endothelial Cells (HCAEC)

500,000 proliferating cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human coronary artery
Morphology:
( endothelial)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human coronary artery

packaging

pkg of 500,000 cells

morphology

( endothelial)

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

room temp

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Application

Primary Human Coronary Artery Endothelial Cells (HCAEC) are isolated from the coronary arteries (the right and the left coronary artery, including the anterior descending and the circumflex branches) from a single donor and are provided in a proliferating format. They are crucially involved in the regulation of coronary blood flow and cardiac functions and are consequently useful for in vitro studies of diseases such as thrombosis, atherosclerosis, and hypertension. HCAEC are also suitable for studying functional vasodilators, such as prostacyclin, bradykinin, or nitric oxide.

Biochem/physiol Actions

Artery

Preparation Note

Click here for more information.

Analysis Note

Rigid quality control tests are performed for each lot of Endothelial Cells from large vessels. They are tested for cell morphology, adherence rate, and cell viability. Flow cytometric analyses for cell-type specific markers, e.g. von Willebrand Factor (vWF) and CD31, as well as Dil-Ac-LDL uptake assays are also carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Other Notes

Recommended Plating Density: 5000 - 10000 cells per cm2Passage After Thawing: P3Tested Markers: von Willebrand Factor (vWF) positive, CD31 positive, Dil-Ac-LDL uptake positiveGuaranteed population doublings: >15
Recommended Primary Cell Culture Media:Link

Disclaimer

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-14022C-12272C-12221
morphology

( endothelial)

morphology

(endothelial)

morphology

( endothelial)

morphology

( endothelial)

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

biological source

human coronary artery

biological source

human coronary artery

biological source

human aorta (thoracic and abdominal)

biological source

human coronary artery

storage temp.

room temp

storage temp.

−20°C

storage temp.

room temp

storage temp.

−196°C

shipped in

ambient

shipped in

dry ice

shipped in

ambient

shipped in

dry ice

packaging

pkg of 500,000 cells

packaging

pkg of 1,000,000 cells

packaging

pkg of 500,000 cells

packaging

pkg of 500,000 cells


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters